Order Entry
ContactUsLinkComponent
VWR* Big Digit Solar-Powered Calculator
VWR® Big-Digit Solar-Powered Calculator
Catalog #: 15551-118
Supplier:  VWR International
VWR* Big Digit Solar-Powered Calculator
VWR® Big-Digit Solar-Powered Calculator
Catalog #: 15551-118
Supplier:  VWR International

Specifications

  • Description:
    VWR® solarcalculator
  • Cat. no.:
    15551-118
  • Dimensions:
    11.4×1×0.6 cm (41/2×23/8×1/4")

Specifications

About this item

Oversized digit calculator is solar-powered, battery-saving, and economical. Calculator performs all arithmetic operations and features memory, percent, and floating decimal point.

  • Easy-to-read display
  • Memory features and arithmetic operations
  • 1.3 cm (0.5") high digits and oversized keys allow for easy typing and viewing

Weight: 57 g (2 oz.)

Ordering information: Supplied with carrying case.
Customers Who Bought This Also Bought