ContactUsLinkComponent
Phase Hemacytometers
Phase Hemacytometer, Hausser Scientific
Catalog #: 15170-263
Supplier:  Hausser
Phase Hemacytometers
Phase Hemacytometer, Hausser Scientific
Catalog #: 15170-263
Supplier:  Hausser
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Phase Hemacytometer
  • Cat. no.:
    15170-263
  • Supplier No.:
    3200
Specifications
About this item
Phase hemacytometer is used with a phase microscope for accurately counting red and white blood cells, platelets, dust/yeast clusters and specimens in spinal and other body fluids
Triple dividing lines show clearly which cells lie within the counting area. A “V” cut into the plateau makes filling easy and reduces the possibility of overflow into the moat. Each plateau features enhanced Neubauer rulings 0.1mm below the cover glass.
Ordering information: Supplied with two reusable cover glasses.
Customers Who Bought This Also Bought