VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Catalog #: 14672-608
Supplier: VWR International
Specifications
- Description:Disposable Controlled Drop Pasteur Pipet
- Cat. no.:14672-608
- Overall Length:14.6 cm (5³/₄")
Specifications
About this item
VWR® Disposable Pasteur Pipettes are ideally suited for use in both clinical and industrial laboratories.
- Made up of borosilicate glass 7.0
- Ideal for both clinical and industrial use
- Supplied in convenient USE-Packs® packaging
Disposable Pasteur pipettes provide a convenient and time-efficient means for inoculating culture media and transferring samples and reagents in both clinical and industrial laboratories. No cotton plug. Pipette body Ext.Ø: 7 mm. Capillary stem Ext.Ø: approximately 1.5 mm. Controlled drop pipette (14672-608) delivers 22 to 28 drops per ml.
Packaging: Supplied in convenient USE-Packs® packages, each containing 200 pipettes.
Customers Who Bought This Also Bought