Straight-Sided Jars, Clear, Wheaton®, DWK Life Sciences
Catalog #: 14235-836
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:89-400
- Capacity:1 L (33.8 oz.)
- Ø:95 mm
- Height:170 mm
- Supplier No.:W216912
Specifications
About this item
Wheaton Straight Sided jars are manufactured from clear soda lime glass that conforms to USP Type III requirements.
- Jars are ideal for soil sampling and environmental applications
The wide mouth design with straight sides easily accommodates large solid samples, and allows for complete removal of contents.
A partitioned carton protects bottles from scratches during shipping, and becomes useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or bulk packs without caps. Extra screw caps can be purchased separately.
Customers Who Bought This Also Bought