Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 14231-116
Supplier: Thermo Fisher Scientific
Specifications
- Volume:0.5 - 200 µl
- Packed:Refill Kit: 20 × 96/Refill Tray
- Tip Style:Finntip™ 250 Universal
- Supplier No.:9400267
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought