Finntip™ Pipette Tips, Thermo Scientific
Catalog #: 14224-022
Supplier: Thermo Fisher Scientific
Specifications
- Volume:0.2 - 50 μL
- Packed:10×384/Rack
- Cat. no.:14224-022
- Tip Style:Finntip™ 50
- Supplier No.:9400370
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought