ContactUsLinkComponent
Stirrer Paddle
Orion™ Automatic Stirrer Probe and Paddle, Thermo Scientific
Catalog #: 14223-214
Stirrer Paddle
Orion™ Automatic Stirrer Probe and Paddle, Thermo Scientific
Catalog #: 14223-214
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Dimensions:
    1.5×1.5×1.5 cm
  • Description:
    Orion™ Star Series Stirrer Replacement Paddles for the Orion™ Star Series Stirrer Probe
  • Supplier No.:
    096021
Specifications
About this item
Use the Orion™ Star Stirrer Probe with Orion™ benchtop meters to stir solutions for better accuracy without the difficulty of using a stir plate and bar.
Use the Orion™ Star Stirrer Probe with Orion™ Versa Star™ meters, Orion™ Star A210 series benchtop meters, and legacy Orion™ Star series benchtop meters to stir standards and samples for better measurement accuracy without the difficulty of separate stir plate and bars. The Orion™ Star Stirrer Probe is powered and controlled by the meter with five optimized stirring speeds. The device fits in the electrode stand for easy movement in and out of samples and can be cleaned simply by rinsing. Paddles can be replaced to extend the life of Orion™ Star Stirrer Probes.
Customers Who Bought This Also Bought