Finntip Stepper Tips, Thermo Scientific
Catalog #: 14222-046
Supplier: Thermo Fisher Scientific
Specifications
- Volume:100 µl, 200 µl, 300 µl, 400 µl, 500 µl
- Sterile:+
- Tip Style:Finntip Stepper 5.0 mL
- Supplier No.:9404203
Specifications
About this item
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought