VWR® Autoclavable Biohazard Bags, Double Thick
Catalog #: 14220-084
Supplier: VWR International
Small Business Enterprise
Specifications
- W×L:610×762 mm (24×30")
- Color:red
- Cat. no.:14220-084
Specifications
About this item
Made of high-strength, highly temperature-resistant 3 mm polypropylene plastic that is autoclavable to 135 °C (275 °F).
- Temperature-resistant polypropylene
- Autoclavable
- Biohazard warning label
Heavy-duty bags offer extra puncture resistance and high impact strength. Clearly labeled with proper biohazard warnings. Bright red color for quick identification.
Ordering information: Bags are supplied with 20.3 cm (8") twist ties.
Customers Who Bought This Also Bought