VWR® Autoclavable Bags, 1.5 mil
Catalog #: 14220-024
Supplier: VWR International
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- W×L:610×914 mm (24×36")
- Color:clear, plain
Specifications
About this item
Autoclave bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
- Bags are available clear or red with proper biohazard warnings, or clear without label
- Rated for use at 135 °C (275 °F)
High-strength, highly temperature-resistant polypropylene construction allows perfect adaptability to standard laboratory disposal routines, including steam autoclaving.
Ordering information: Twist ties included.
Customers Who Bought This Also Bought