Order Entry
ContactUsLinkComponent
pH "All in One" Buffer Kit
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
Catalog #: 14218-368
CAS Number:  
pH "All in One" Buffer Kit
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
Catalog #: 14218-368
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Standard All-in-One pH buffer kit
  • Cat. no.:
    14218-368
  • Supplier No.:
    910199

Specifications

About this item

This pH buffer kit has colour coded buffers, storage solution and storage bottles.

They are manufactured under ISO 9000 quality standards. The kit includes 1 pint (475 ml) bottle each of pH 4.00 buffer, pH 7.00 buffer, pH 10.01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.

pH buffers with temperature tables on the labels are avilable for accurate calibrations and NIST™ traceability.

Customers Who Bought This Also Bought