Order Entry
ContactUsLinkComponent
CYLINDER GRADUATED TD 10ML
SP Wilmad-LabGlass Graduated Cylinders, with Detachable Plastic Base and Guard, TD, SP Industries
Catalog #: 14212-512
Supplier:  SP Industries
CYLINDER GRADUATED TD 10ML
SP Wilmad-LabGlass Graduated Cylinders, with Detachable Plastic Base and Guard, TD, SP Industries
Catalog #: 14212-512
Supplier:  SP Industries

Specifications

  • Capacity:
    10 ml
  • Graduation Interval:
    0.2 ml
  • Form:
    Short
  • Cat. no.:
    14212-512
  • Supplier No.:
    LG-5517-100

Specifications

About this item

These borosilicate glass graduated cylinders are calibrated "to deliver" (TD).

  • Cylinders feature a single metric scale
  • Supplied with glass cylinder, detachable plastic base, and plastic collar
  • See LG-5517A and LG-5518 for replacement parts
Customers Who Bought This Also Bought