SP Wilmad-LabGlass Gay-Lussac Specific Gravity Bottles, SP Industries
Catalog #: 14210-660
Supplier: SP Industries
Specifications
- Capacity:10 ml
- Cat. no.:14210-660
- ST Stopper No.:10/18
- Supplier No.:LG-3530-104
Specifications
About this item
These unadjusted borosilicate glass specific gravity bottles determine the density of liquid samples
They feature capillary stoppers, and ground Standard Taper joints to reduce evaporation.
Customers Who Bought This Also Bought