Order Entry
ContactUsLinkComponent
Medium Waste Container
BETA-GARD™ Waste Containers, IBI Scientific
Catalog #: 13500-536
Supplier:  IBI Scientific
Medium Waste Container
BETA-GARD™ Waste Containers, IBI Scientific
Catalog #: 13500-536
Supplier:  IBI Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Dimensions:
    30.5W x 22.9D x 40.6H cm (12 x 9 x 16")
  • Size:
    Medium
  • Cat. no.:
    13500-536
  • Supplier No.:
    WH-160

Specifications

About this item

Containers for the disposal of radioactive waste can also be used with disposable NRC-approved waste bags

Containers for the disposal of radioactive waste can also be used with disposable NRC-approved waste bags

Constructed of durable 13mm (1/2") cast acrylic for maximum protection. Available with hinged or flip-top lids. Container IBWH-280-M has a handle. Containers with flip-top lids have a 2cm hole for disposal of contaminated pipets.

Customers Who Bought This Also Bought