DuPont™ Tyvek® Coveralls with Attached Hood, Magid Glove
Catalog #: 13450-166
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Clothing Size:L
- Garment application:Industrial,Laboratory
- Color:White
- Description:DuPont™ Tyvek® Coveralls with Attached Hood
- Seam type:Serged
- Cat. no.:13450-166
- Supplier No.:CVCH11L
Specifications
About this item
Disposable coveralls are made of Tyvek®, a tough polyolefin material which provides the best balance of protection, durability, and comfort.
- Zippered front
- Remains strong, wet or dry
- Elastic border at wrists and ankles
- Comfortable and more tailored fit
These lightweight overalls feature a great range of movement while stretching and bending, improved mobility, a tailored fit, and reinforcement in high stress areas.
Coveralls are ideal for use against such non-hazardous materials as dirt and grime, animal waste, paints, oil and grease, lubricants, or hazardous materials including fertilizer, pesticides, asbestos, lead, chromium, mold, fiberglass, and radioactive particles.
Customers Who Bought This Also Bought