Magid® EconoWear® Disposable Laboratory Coats, Magid
Catalog #: 13450-164
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Clothing Size:3XL
- Garment application:Laboratory
- Pockets:With pockets
- Color:White
- Description:EconoWear® Disposable Lab Coat
- Cat. no.:13450-164
- Supplier No.:CC1114XXXL
Specifications
About this item
EconoWear® disposable lab coats are made of Tyvek, a tough polyolefin material, which provides the optimal balance of protection, durability and comfort.
- Elastic wrists
- Collar with snap front
- Greater range of movement while stretching and bending
These coveralls feature a great range of movement while stretching and bending, improved mobility, a tailored fit, and reinforcement in high stress areas. Elastic at wrist openings creates a comfortable seal against contaminants. For use against such non-hazardous materials as dirt and grime, animal waste, paints, oil and grease, lubricants, or hazardous materials including: fertilzer, pesticides, asbestos, lead, chromium, mold, fiberglass, and radioactive particles.
Customers Who Bought This Also Bought