ContactUsLinkComponent
VWR* Modular Heating Block
VWR® Modular Heating Block for Titer Plates
Catalog #: 13259-295
Supplier:  VWR International
Product available on GSA Advantage®
VWR* Modular Heating Block
VWR® Modular Heating Block for Titer Plates
Catalog #: 13259-295
Supplier:  VWR International
Product available on GSA Advantage®
Specifications
  • Description:
    Block, 96 Test Well Plate, Titer Plate
  • Cat. no.:
    13259-295
  • L×W×H:
    15.1 x 9.5 x 5.7 cm (6 x 33/4 x 21/4")
  • Block Size:
    Double
Specifications
About this item
Heating block is ideal for use with 96-well or 384-well titer plates.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
It features a recessed well for better stability and a flat surface appropriate for flat or round-bottom plates. Double block size fits only 2, 4, or 6 block heaters.
Customers Who Bought This Also Bought