OIML Class F1 Precision Weights, Troemner
Catalog #: 12766-356
Supplier: Troemner
Product available on GSA Advantage®
Specifications
- Cat. no.:12766-356
- Certification:Statement of Accuracy
- Weight:200 mg
- Supplier No.:7528-F1
Specifications
About this item
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are two-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.
Customers Who Bought This Also Bought