VWR® Signature™ Glass Beads, Silica
Catalog #: 12621-154
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Ø:400 µm
- Description:Zirconia molecular biology grade grinding beads, 400 µm
- Environmentally Preferable:Waste Reducing
- Environmental certification:RoHS
- Cat. no.:12621-154
Specifications
About this item
Molecular biology grade silica beads, grinding media for VWR® Signature™ Pulsing Vortex Mixers.
- Treated to be DNase/RNase-free
- Useful for preparing samples for molecular biology applications
Customers Who Bought This Also Bought