VWR® Modular Heating Blocks for Vials
Catalog #: 12621-126
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:15 mm Vial Block
- No. of wells:20
- Cat. no.:12621-126
- For Vials:Ext.Ø 15 mm
- L×W×H:9.5 x 7.6 x 5.1 cm (33/4 x 3 x 2")
- For Use With:Vials
Specifications
About this item
Designed for sample/serum and scintillation vials, these units are constructed from a solid anodized aluminum block.
- Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.
Customers Who Bought This Also Bought