VWR® Aluminum Micro Weighing Dishes
Catalog #: 12577-060
Supplier: VWR International
Specifications
- Capacity:0.035 ml
- Dimensions:6dia. x 2.5D mm
- Description:Round Weighing Dishes
- Cat. no.:12577-060
Specifications
About this item
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
- Easy to use
- Compatible with most commercially available analyzers and microbalances
- For use in a variety of applications
Our micro weighing boats are compatible with most commercially available analyzers and microbalances. Packaged in easy-to-use plastic containers for storage and dispensing.
Customers Who Bought This Also Bought