Logger Calibration Software
Catalog #: 12444-044
Supplier: Dickson
Customers Who Bought This Also Bought
Ethylene glycol ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$401.45 to $1,217.04
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
$84.04 to $651.48
Methanol-[D₄] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
$374.70
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer, Thermo Scientific
Supplier: Thermo Scientific
$628.97