Order Entry
ContactUsLinkComponent
250cc Natural HDPE Oblong
Oblong Bottles, High-Density Polyethylene, Wide Mouth
Catalog #: 11311-598
Supplier:  Qorpak
Small Business Enterprise
250cc Natural HDPE Oblong
Oblong Bottles, High-Density Polyethylene, Wide Mouth
Catalog #: 11311-598
Supplier:  Qorpak
Small Business Enterprise

Specifications

  • Cap Size:
    43-400
  • Cap Type:
    PE foam lined white polypropylene cap
  • Capacity:
    250 ml (8.5 oz.)
  • Cat. no.:
    11311-598
  • Supplier No.:
    PLC-07195

Specifications

About this item

Natural HDPE wide mouth Oblong bottles offer a space saving design for the workbench.

  • Bottles feature a wide mouth opening, ideal for sampling solids and powders
  • HDPE construction provides an excellent moisture barrier and reliable impact resistance

They are ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping.

Ordering information: Bottles 16196-299, 16196-302, and 16125-976 are supplied pre-cleaned for volatiles. Bottles 11311-580, 11311-596, 11311-598, and 11311-600 are supplied Vacuumed and Ionized.
Customers Who Bought This Also Bought