Finntip™ Multistepper Pipette Tips, Thermo Scientific
Catalog #: 11310-847
Supplier: Thermo Fisher Scientific
Specifications
- Volume:1500 µl
- Packed:10×96/Rack
- Filter:+
- Sterile:+
- Tip Style:Finntip Multistepper
- Supplier No.:94052450
Specifications
About this item
Finntip Multistepper Pipette Tips optimize the performance of Thermo Scientific Finnpipette multistepper pipetters.
- Accommodates standard 9 mm microplate spacing
- Sterile or non-sterile versions
They are specifically designed for use with the Thermo Scientific Finnpipette Multistepper Pipetter. This tip has a nominal volume of 1500 μl and accommodates standard 9 mm microplate spacing. Finntip multistepper tips are available in racks of 96 tips, in both sterile and non-sterile versions.
Customers Who Bought This Also Bought