SP Bel-Art Biohazard Sharp Object Pouch and Stand, Bel-Art Products, a part of SP
Catalog #: 11215-836
Supplier: Bel-Art Products, a Part of SP
Specifications
- Dimensions:14 x 33 cm (51/2 x 13")
- Description:Pouch
- Cat. no.:11215-836
- Supplier No.:132340000
Specifications
About this item
This solid paperboard pouch is 10 mil thick and offers protection against sharp-edged objects prior to and during disposal.
- Protects against sharp-edged objects for disposal
- Top flaps close and lock in place
- Epoxy-coated welded steel wire stand provides easy, upright access
This solid paperboard pouch is 10 mil thick and offers protection against sharp-edged objects prior to and during disposal.
It remains open in an oval shape by folding the bottom flaps. After filling, the top flaps close and lock in place for safe disposal. Printed with biohazard symbol. Poxygrid® Safety Pouch Stand (11215-847), made of welded steel wire with long-lasting epoxy coating, keeps pouch standing upright for easy access.
Customers Who Bought This Also Bought