Order Entry
ContactUsLinkComponent
Terminal* Pipet Keeper*
Terminal® Pipet Keeper™ Disposal Container, Whitney Products
Catalog #: 11214-715
Supplier:  Whitney Products
Terminal* Pipet Keeper*
Terminal® Pipet Keeper™ Disposal Container, Whitney Products
Catalog #: 11214-715
Supplier:  Whitney Products

Specifications

  • Dimensions:
    39.4L×7.6W×7.6H cm (15¹/₂×3×3")
  • Cat. no.:
    11214-715
  • Supplier No.:
    BH2006

Specifications

About this item

For the collection of biohazard waste at the point of generation.

For the collection of biohazard waste at the point of generation. Holds pipets, swabs, and other objects up to 38.1cm (15") long. Constructed of sturdy, plastic-lined paperboard for leak resistance. Autoclavable container has a reclosable flap. Low profile prevents tipping.

  • Constructed of sturdy, plastic-lined paperboard for leak resistance
  • Autoclavable container has a reclosable flap
  • Low profile prevents tipping

Holds pipets, swabs, and other objects up to 38.1cm (15") long.

Customers Who Bought This Also Bought