Terminal® Biohazard Benchtop Keeper™ Containers, Whitney Products
Catalog #: 11214-708
Supplier: Whitney Products
Specifications
- Capacity:1.4 L (47.3 oz.)
- Dimensions:10.2L×10.2W×17.8H cm (4×4×7)"
- Cat. no.:11214-708
- Supplier No.:BH2002
Specifications
About this item
Use large or small containers for the collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides.
Use large or small containers for the collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides. Constructed of sturdy, plastic-lined paperboard. Disposable containers are leak-resistant and autoclavable. Self-locking top for safe closure.
- For collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides
- Sturdy, plastic-lined paperboard construction
- Leak-resistant and autoclavable
Constructed of sturdy, plastic-lined paperboard. Disposable containers are leak-resistant and autoclavable. Self-locking top for safe closure.
Customers Who Bought This Also Bought