Laboratory Coats, PP, Cellucap
Catalog #: 10833-004
Supplier: Cellucap-Melco
Specifications
- Clothing Size:L
- Garment application:Laboratory
- Color:White
- Cat. no.:10833-004
- Supplier No.:3330W-LRG
Specifications
About this item
A heavier polypropylene alternative for splash protection from non-hazardous liquids and dry particulates.
- Snap front
- Left breast pocket
- 2 hip pockets
- Knit cuffs and collar
Customers Who Bought This Also Bought