Alliance Contamination Control Coveralls, HPK Industries
Catalog #: 10832-250
Supplier: HPK Industries
Specifications
- Clothing Size:XL
- Garment application:Cleanroom,Laboratory
- Cat. no.:10832-250
- Supplier No.:150-XL
Specifications
About this item
An extremely smooth outer fabric surface lowers particle cling and build-up in cleanroom applications
Fabric is wrinkle-free and highly durable. Built-in “cooling system” dissipates heat and moisture for high worker comfort. Elastic wrists and ankles. Hood and boot covers (shown) not included.
Individually packaged in a sealed bag.
Bound seam construction. Coverall material acts as a liquidproof barrier. Unique antistatic system assures static dissipation in sensitive cleanroom work areas.
Customers Who Bought This Also Bought