Anti-MED27 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: MED27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77831-405
- 77831-404
- Antigen symbol:
- MED27
- MED27
- Antigen name:
- Mediator of RNA polymerase II transcription subunit 27 protein (39-108AA)
- Mediator of RNA polymerase II transcription subunit 27 protein (39-108AA)
- Antigen synonyms:
- Cofactor required for Sp1 transcriptional activation subunit 8 (34kD)|Cofactor required for Sp1 transcriptional activation subunit 8 34kDa|Cofactor required for Sp1 transcriptional activation subunit 8|CRAP 34|CRAP34|CRSP 34|CRSP 8|CRSP complex subunit 8|CRSP34|CRSP8|MED 27|Med27|MED27_HUMAN|Mediator complex subunit 27|Mediator of RNA polymerase II transcription subunit 27|MGC11274|P37 TRAP/SMCC/PC2 subunit|Transcriptional coactivator CRSP34|TRAP 37|TRAP37
- Cofactor required for Sp1 transcriptional activation subunit 8 (34kD)|Cofactor required for Sp1 transcriptional activation subunit 8 34kDa|Cofactor required for Sp1 transcriptional activation subunit 8|CRAP 34|CRAP34|CRSP 34|CRSP 8|CRSP complex subunit 8|CRSP34|CRSP8|MED 27|Med27|MED27_HUMAN|Mediator complex subunit 27|Mediator of RNA polymerase II transcription subunit 27|MGC11274|P37 TRAP/SMCC/PC2 subunit|Transcriptional coactivator CRSP34|TRAP 37|TRAP37
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P2C8
- Q6P2C8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together