Anti-CEACAM8 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: CEACAM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77812-111
- 77812-110
- Antigen symbol:
- CEACAM8
- CEACAM8
- Antigen name:
- Fusion protein of CEACAM8
- Fusion protein of CEACAM8
- Antigen synonyms:
- Carcinoembryonic antigen CGM6|Carcinoembryonic antigen gene family member 6|Carcinoembryonic antigen related cell adhesion molecule 8|Carcinoembryonic antigen-related cell adhesion molecule 8|CD 66b|CD 67|CD66b|CD66b antigen|CD67|CD67 antigen|CEACAM 8|CEAM8_HUMAN|CGM 6|CGM6|NCA 95|NCA95|Non-specific cross-reacting antigen NCA-95|Nonspecific cross reacting antigen NCA 95|Nonspecific cross reacting antigen NCA95
- Carcinoembryonic antigen CGM6|Carcinoembryonic antigen gene family member 6|Carcinoembryonic antigen related cell adhesion molecule 8|Carcinoembryonic antigen-related cell adhesion molecule 8|CD 66b|CD 67|CD66b|CD66b antigen|CD67|CD67 antigen|CEACAM 8|CEAM8_HUMAN|CGM 6|CGM6|NCA 95|NCA95|Non-specific cross-reacting antigen NCA-95|Nonspecific cross reacting antigen NCA 95|Nonspecific cross reacting antigen NCA95
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31997
- P31997
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together