Anti-USP48 Rabbit Polyclonal Antibody
Catalog # 77810-252
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of USP48.
- Antigen symbol:USP48
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q86UV5
- Antigen synonyms:AAH67261|Deubiquitinating enzyme 48|DKFZp762M1713|EC 3.1.2.15|Kiaa4202|MGC132556|MGC14879|RAP1GA1|Synaptic ubiquitin specific protease|synUSP|Ubiquitin carboxyl terminal hydrolase 48|Ubiquitin carboxyl-terminal hydrolase 48|Ubiquitin specific peptidase 48|Ubiquitin specific processing protease 48|Ubiquitin specific protease 31|Ubiquitin specific protease 48|Ubiquitin thioesterase 48|Ubiquitin thiolesterase 48|Ubiquitin-specific-processing protease 48|UBP48_HUMAN|USP31|USP48
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: USP48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat