Anti-YEATS4 Rabbit Polyclonal Antibody
Catalog # 77809-303
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Synthesized peptide derived from the N-terminal region of GAS41.
- Antigen symbol:YEATS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O95619
- Antigen synonyms:4930573H17Rik|B230215M10Rik|GAS 41|Gas41|glioma amplified sequence 41|Glioma-amplified sequence 41|glioma-amplified sequence-41|gliomaamplified sequence 41|gliomaamplified sequence41|NUBI 1|NuBI-1|NuBI1|NuMA binding protein 1|NuMA-binding protein 1|YAF9|YEATS domain containing 4|YEATS domain containing4|YEATS domain-containing protein 4|YEATS4|YETS4_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000
Type:
Antigen: YEATS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse