Anti-UCHL5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: UCHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77820-217
- 77820-216
- Antigen symbol:
- UCHL5
- UCHL5
- Antigen name:
- Fusion protein of UCHL5
- Fusion protein of UCHL5
- Antigen synonyms:
- UCH37|AD-019|CGI 70|INO80 complex subunit R|INO80R|Ubiquitin C terminal hydrolase L5|Ubiquitin C terminal hydrolase UCH37|Ubiquitin C-terminal hydrolase UCH37|Ubiquitin carboxyl terminal esterase L5|Ubiquitin carboxyl terminal hydrolase isozyme L5|Ubiquitin carboxyl terminal hydrolase L5|Ubiquitin carboxyl-terminal hydrolase isozyme L5|Ubiquitin thioesterase L5|Ubiquitin thiolesterase L5|UCH L5|UCH-L5|UCHL5|UCHL5_HUMAN
- UCH37|AD-019|CGI 70|INO80 complex subunit R|INO80R|Ubiquitin C terminal hydrolase L5|Ubiquitin C terminal hydrolase UCH37|Ubiquitin C-terminal hydrolase UCH37|Ubiquitin carboxyl terminal esterase L5|Ubiquitin carboxyl terminal hydrolase isozyme L5|Ubiquitin carboxyl terminal hydrolase L5|Ubiquitin carboxyl-terminal hydrolase isozyme L5|Ubiquitin thioesterase L5|Ubiquitin thiolesterase L5|UCH L5|UCH-L5|UCHL5|UCHL5_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5K5
- Q9Y5K5
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together