Anti-KDM5A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:300
Type:
Antigen: KDM5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77832-924
- 77832-923
- Antigen symbol:
- KDM5A
- KDM5A
- Antigen name:
- Synthetic peptide of KDM5A
- Synthetic peptide of KDM5A
- Antigen synonyms:
- Histone demethylase JARID1A|JARID1A|Jumonji/ARID domain containing protein 1A|Jumonji/ARID domain-containing protein 1A|Kdm5a|KDM5A_HUMAN|Lysine-specific demethylase 5A|RBBP-2|RBBP2|RBP2|Retinoblastoma binding protein 2|Retinoblastoma-binding protein 2
- Histone demethylase JARID1A|JARID1A|Jumonji/ARID domain containing protein 1A|Jumonji/ARID domain-containing protein 1A|Kdm5a|KDM5A_HUMAN|Lysine-specific demethylase 5A|RBBP-2|RBBP2|RBP2|Retinoblastoma binding protein 2|Retinoblastoma-binding protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29375
- P29375
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together