Anti-RCBTB2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RCBTB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77796-913
- 77796-912
- Antigen symbol:
- RCBTB2
- RCBTB2
- Antigen name:
- RCC1 and BTB domain-containing protein 2 protein (1-200AA)
- RCC1 and BTB domain-containing protein 2 protein (1-200AA)
- Antigen synonyms:
- CHC1 L| CHC1L| Chromosome condensation 1 like| RCBT2| RCBT2_HUMAN| Rcbtb2| RCC1 and BTB domain containing protein 2| RCC1 and BTB domain-containing protein 2| RCC1 like G exchanging factor| RCC1 like G exchanging factor RLG| Regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2| Regulator of chromosome condensation (RCC1) and BTB domain containing protein 2| Regulator of chromosome condensation and BTB domain containing protein 2| RLG
- CHC1 L| CHC1L| Chromosome condensation 1 like| RCBT2| RCBT2_HUMAN| Rcbtb2| RCC1 and BTB domain containing protein 2| RCC1 and BTB domain-containing protein 2| RCC1 like G exchanging factor| RCC1 like G exchanging factor RLG| Regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2| Regulator of chromosome condensation (RCC1) and BTB domain containing protein 2| Regulator of chromosome condensation and BTB domain containing protein 2| RLG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95199
- O95199
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together