Anti-PRPF4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77795-570
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:U4/U6 small nuclear ribonucleoprotein Prp4 protein (1-260AA)
- Antigen symbol:PRPF4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O43172
- Antigen synonyms:hPrp4|HPRP4P|PRP4|PRP4 homolog|PRP4 pre mRNA processing factor 4 homolog (yeast)|PRP4/STK/WD splicing factor|PRP4_HUMAN|Prp4p|PRPF4|RP70|SNRNP60|U4/U6 small nuclear ribonucleoprotein Prp4|U4/U6 snRNP 60 kDa protein|WD splicing factor Prp4
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PRPF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human