Chromatography Column, #50 or #80, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These large size chromatography columns have an O-Ring-grooved Duran flat flange at the top for easy access to the column.
- Fabricated from borosilicate glass
The flange is supplied with a silicone O-Ring to make a seal when using 89053-052, 89053-054, or 89053-056 heads with a 89055-110 (4") or 89055-112 (6") quick-release clamp. Both the 4 and 6" int.Ø. columns are tapered to #50 or #80 Ace-Thred at bottom that accepts various adapters.
Ordering information: For replacement silicone O-Rings, order 71000-162 (for 4-inch flange) or 71000-164 (for 6-inch flange). For replacement CAPFE O-Rings, order 89061-166 (for 4-inch flange) or 89061-168 (for 6-inch flange). Column extenders are also available. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought