Column Head, #15 Ace-Thred, Ace Glass
Supplier: Ace Glass
Certificates
About this item
For use with chromatography columns.
Ordering information: Four-inch version requires 89055-110 clamp; six-inch version requires 89055-112 clamp. Clamp is NOT included, and must be ordered separately.
Customers Who Bought This Also Bought