Copper(II) sulfate pentahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: Cupric sulfate pentahydrate,Cupric sulphate pentahydrate,Copper sulphate pentahydrate
Formula:CuSO₄·5H₂O
MW:249.69 g/mol
Density:2.284 g/cm³ (20 °C)
MDL Number:MFCD00149681
CAS Number:7758-99-8
ADR:9,III
Merck Index:13,02682
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 249.69 g/mol
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
REACH:01-2119520566-40
:9
:III
:GHS07
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Calcium:= 0.005 %
- Chloride:= 0.001 %
- Insolubles:= 0.005 %
- Iron:= 0.003 %
- Nickel:= 0.005 %
- Nitrogen Compounds (as N):= 0.002 %
- Potassium:= 0.01 %
- Purity:98.0 - 102.0 %
- Sodium:= 0.02 %
- Meets reagent specifications for testing USP/NF monographs:
- VWR Chemicals BDH®, Copper(II) sulfate pentahydrate 98.0-102.0% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
BDH9312-2.5KG_BSETSE BDH9312-500G_BSETSE
BSE/TSE Certificates
spec_BDH9312.pdf
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought