Potassium hydrogen phthalate 99.95-100.05% ACS, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: Potassium phthalate monobasic,Potassium biphthalate,Potassium Acid Phthalate,KHP,Phthalic Acid Monopotassium Salt
Formula:HOOCC₆H₄COOK
MW:204.22 g/mol
Melting Pt:295…300 °C
Density:1.6362 g/cm³ (20 °C)
MDL Number:MFCD00013070
CAS Number:877-24-7
Merck Index:12,07773
:Potassium hydrogen phthalate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,95-100,05
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 295 to 300 °C|Melting Pt: 295…300 °C
:Flash Pt:
:Flash Pt:
:M.W. 204.22 g/mol
Storage Temperature:Ambient
EINECS:212-889-4
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (dried basis):99.95 - 100.05 %
- Chlorine Compounds:= 0.003 %
- Heavy Metals (as Pb):= 5 ppm
- Insoluble Matter:= 0.005 %
- Iron:= 5 ppm
- pH (0.05M, Water) @25C:4.00 - 4.02
- Sodium:= 0.005 %
- Sulfur Compounds:= 0.002 %
- VWR Chemicals BDH®, Potassium hydrogen phthalate 99.95-100.05% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
BDH9260-125G_BSETSE BDH9260-500G_BSETSE
BSE/TSE Certificates
spec_BDH9260.pdf
Product specification
Customers Who Bought This Also Bought