Magnesium chloride hexahydrate 99.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Formula:MgCl₂·6H₂O
MW:203.3 g/mol
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
MDL Number:MFCD00149781
CAS Number:7791-18-6
Merck Index:13,05684
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-102,0
:Boiling Pt:
:Boiling Pt: 1412 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 117 °C
:Flash Pt:
:Flash Pt:
:M.W. 203.3 g/mol
:Magnesium chloride hexahydrate
Storage Temperature:Ambient
EINECS:232-094-6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Purity:99.0-102.0 %
- Ammonium:≤0.002 %
- Barium:≤0.005 %
- Calcium:≤0.01 %
- Heavy Metals (as Pb):≤0.0005 %
- Insolubles:≤0.005 %
- Iron:≤0.0005 %
- Manganese:≤0.0005 %
- Nitrate:≤0.001 %
- Phosphate:≤0.0005 %
- Potassium:≤0.005 %
- Sodium:≤0.005 %
- Strontium:≤0.005 %
- Sulfate:≤0.002 %
- Meets reagent specifications for testing USP/NF monographs:
- VWR Chemicals BDH®, Magnesium chloride hexahydrate 99.0-102.0% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals F&BFOCUS Volume 5 Issue 1w
Product Literature
BDH9244-12KG_BSETSE BDH9244-2.5KG_BSETSE BDH9244-500G_BSETSE
BSE/TSE Certificates
spec_BDH9244
Product specification
Customers Who Bought This Also Bought