Wire Workstation, Stationary, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Stationary wire workstation units offer 62% laminar flow
24" wide wire shelving workstation top. Four 33" legs and C-frame for additional stability. Weight capacity of up to 800 lbs evenly distributed. All units have standard working height of 33"
Customers Who Bought This Also Bought