Mini Safety Cabinet, Justrite®
Catalog #: 89209-854
Supplier: Justrite
Specifications
- Description:Mini safety cabinet
- Cat. no.:89209-854
- Supplier No.:890200
Specifications
About this item
This safety cabinet offers the smallest footprint for storing small quantities of flammables for cleaning and maintenance at point-of-use applications
- Color: yellow
The cabinet includes three bungee cords to keep contents in place when transporting cabinet on service vehicles, carts or maintenance trikes.
Customers Who Bought This Also Bought