Order Entry
ContactUsLinkComponent
SP Bel-Art Jaw Style Tubing Clamp, Acetal, Bel-Art Products, a part of SP
SP Bel-Art Jaw Style Tubing Clamp, Acetal, Bel-Art Products, a part of SP
Catalog #: 70690-116
CLAMP, ACETAL, TUBING, 12/PKG, 72/CASE
SP Bel-Art Jaw Style Tubing Clamp, Acetal, Bel-Art Products, a part of SP
Catalog #: 70690-116

Specifications

  • Cat. no.:
    70690-116
  • Fits Tubing O.D.:
    Up to 0.82 mm (¹⁄₃₂") wall thickness
  • Supplier No.:
    182180000

Specifications

About this item

Tube clamp is molded of strong, durable plastic with serrated jaws 2.7 cm (1¹⁄₁₆") long.

  • Designed for one-handed operation
  • Strong, durable plastic construction
  • Serrated jaws
  • Autoclavable

Attach at any point on tubing without disconnecting tubing

When not in use, the circular opening (approximately 0.6 cm (¼") diameter) at hinge end allows clamp to remain attached to tubing having 0.6 - 1.0 cm (¼ - ⅜") OD with little to no restriction in flow. Overall length is 5.9 cm (2⁵⁄₁₆"). Autoclavable.

Designed to be operated with one hand, the clamp locks closed and opens when the locking tab is released. For use on tubing up to 1.3 cm (½") O.D. and wall thickness of 0.82 mm (1/32") or thinner.

Customers Who Bought This Also Bought