To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein C-Iii
- Clonality:Polyclonal
- Gene ID:345
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:APOC3
- Conjugation:Biotin
- ELISA:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:APOCIII|HALP2
- Storage buffer:Preservative: 0.02% Sodium Azide. Constituents: 0.15M Sodium chloride, 0.01M PBS. pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4oC short term. For extended storage aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
- Concentration:0.85 mg/ml
- Immunogen:Purified human Apolipoprotein CIII.
- Tested applications:ELISA, WB
- Purification:Immunogen affinity purified
- Cat. no.:89363-414
- Supplier No.:GTX41024
Specifications
About this item
Biotin-conjugated Goat polyclonal to Apolipoprotein CIII (Biotin)
ELISA: 1/8000 - 1/30000. IA: Use at an assay dependent dilution. WB: 1/8000 - 1/30000. Predicted molecular weight: 11 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: APOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human