To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein B
- Clonality:Monoclonal
- Clone:6G1
- Gene ID:6285
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:S100B
- Conjugation:Unconjugated
- Size:500 μg
- Western blot:Yes
- Antigen synonyms:S100beta|protein S100-B|S-100 calcium-binding protein|beta neural|NEF|S100-B|S100|S-100 protein subunit beta|S100 calcium-binding protein|beta chain
- Storage buffer:Preservative: 0.1% Sodium Azide. Constituents: PBS. pH 7.4
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:9.6 mg/ml
- Immunogen:Purified S100 protein from human brain.
- Tested applications:Affinity Purification, Immuno Assay
- Purification:Protein A purified
- Cat. no.:89352-824
- Supplier No.:GTX40327
Specifications
About this item
Mouse monoclonal [6G1] to S100 beta
AP: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 11 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: S100B
Clonality: Monoclonal
Clone: 6G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human