To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Alpha 1, Skeletal Muscle
- Clonality:Polyclonal
- Gene ID:60
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ACTA1
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Storage buffer:Tris-citrate/phosphate containing 0.09% sodium azide, pH 7-8.
- Storage temperature:Store at 4°C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen corresponding to the N-terminus of human Beta Actin (Swiss-Prot entry P60709).
- Tested applications:IP, WB
- Purification:Immunogen affinity purified
- Cat. no.:89352-712
- Supplier No.:GTX30632
Specifications
About this item
Rabbit polyclonal antibody to beta Actin
WB: 1:5,000-1:15,000. A 40 kDa band is detected in HeLa whole cell lysate and mouse NIH3T3 cells.
Type: Primary
Antigen: ACT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human