To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 2
- Clonality:Monoclonal
- Clone:JA22
- Gene ID:10370
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CITED2
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Format:Ascites
- Antigen synonyms:P35SRJ|MRG1|Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2
- Storage temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Concentration:22.4 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested applications:WB
- Purification:Ascites
- Cat. no.:89362-906
- Supplier No.:GTX30290
Specifications
About this item
Anti - CITED2 monoclonal
WB: 1/500. Detects a band of approximately 29 kDa (predicted molecular weight: 28 kDa).
Type: Primary
Antigen: CITED2
Clonality: Monoclonal
Clone: JA22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: