To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Telomerase Reverse Transcriptase
- Clonality:Monoclonal
- Clone:2D8
- Gene ID:7015
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Antigen symbol:TERT
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:TRT|Telomerase catalytic subunit|Telomerase-associated protein 2|Telomerase reverse transcriptase|EST2|HEST2|TP2|TCS1
- Storage buffer:Preservative: 0.1% Sodium Azide; Constituents: PBS, Raw Ascites
- Storage temperature:Store at –20 degrees C. Avoid repeated freeze-thaw cycles.
- Concentration:3.3 mg/ml
- Immunogen:Recombinant full length protein (Human), expressed in insect cells.
- Tested applications:ICC/IF, WB
- Purification:Ascites
- Cat. no.:89361-626
- Supplier No.:GTX25405
Specifications
About this item
Mouse monoclonal [2D8] to TERT
IF: Use at an assay dependent dilution. WB: Use at a dilution of 1/500 - 1/1000. Detects a band of approximately 127 kDa (predicted molecular weight: 135 kDa). May detect an additional background band of approximately 110 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TERT
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human